ContactUsLinkComponent
ACTINOSPHAERIUM 35 STUDENTS
Ward's® Live Actinosphaerium Sp. Culture
Catalog # 470176-356
Supplier:  Ward's Science
ACTINOSPHAERIUM 35 STUDENTS
Ward's® Live Actinosphaerium Sp. Culture
Catalog # 470176-356
Supplier:  Ward's Science
Specifications
  • Description:
    Actinosphaerium (small)
  • Cat. no.:
    470176-356
  • Media:
    Soil water medium
  • No. of Students:
    30
  • Storage Conditions:
    Room temp, loose cap
  • Packaging:
    Jar
  • Supplier No.:
    WARD470176-356
Specifications
About this item
With stiff 'arms' radiating in all directions, these microscopic organisms look like sea urchins
  • Large protist
  • Excellent example of a Heliozoan, or "sun animal".
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.
Customers Who Bought This Also Bought