Ward's® Live Green Hydra (Chlorohydra viridissima)
Catalog # 470176-322
Supplier: Ward's Science
Specifications
- Description:Green Hydra Culture (Small)
- Storage Conditions:Refrigerator, loose cap
- No. of Students:30
- Cat. no.:470176-322
- Media:Dechlorinated water
- Supplier No.:WARD470176-322
Specifications
About this item
Hydra are tiny animals closely related to jellyfish. They can be carnivorous as well as photosyntheic due to their symbiotic relationship with green algae Zoochlorellae.
- Watch the feeding reflex in real-time
- Observe the reproductive process of budding
- Green hydra are smaller than the strictly carnivorous, Brown Hydra (also available)
Hydra have a sack-shaped body with a ring of tentacles around the mouth at the top. They use those tentacles, covered with paralyzing nematocysts, to trap small cruscaceans, stun them and bring them into the mouth where they are digested.
Ordering information: These products contain living or perishable materials.
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Frequently Bought Together