Didinium
# de catalogue 470176-266
Fournisseur: Ward's Science
Spécifications
- Description:Didinium (Small)
- No. of Students:30
- Cat. no.:470176-266
- Packaging:Jar
- Media:Hay culture medium
- Supplier No.:WARD470176-266
Spécifications
A propos de cet article
Predatory. Feeds on Paramecium caudatum. Hay medium.
Characterized by cilia; nearly all possess two types of nuclei.
Customers Who Bought This Also Bought