Ward's® Live Ramshorn Snail
# de catalogue 470176-258
Fournisseur: WATERSCAPES AQUATIC NURSERY
Spécifications
- Description:Ramshorn Snail
- Cat. no.:470176-258
- Body Size:¼ - ½"
- Supplier No.:WARD470176-258
Spécifications
A propos de cet article
Planorbids, mixed genera. Compare these attractive, spiral-shelled, herbivorous freshwater specimens with other snails. They will eat aquarium plants
- Herbivorous freshwater snails that will interact well with other snails.
- Will help clean your aquarium by eating debris and algae
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought