Ward's® Freshwater Copepods
# de catalogue 470176-228
Fournisseur: Ward's Science
Spécifications
- Description:Copepods, Freshwater
- No. of Students:35
- Cat. no.:470176-228
- Supplier No.:WARD470176-228
Spécifications
A propos de cet article
The culture supplied includes copepods in various stages of development.
Cyclops or similar genus. Copepods molt numerous times in the nauplius and copepodid stages before reaching adulthood.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought