Order Entry
ContactUsLinkComponent
COTTON ABSORBENT NON-STERILE 1LB ROLL
Cotton Roll
Catalog # 470161-446
COTTON ABSORBENT NON-STERILE 1LB ROLL
Cotton Roll
Catalog # 470161-446

Specifications

  • Description:
    Cotton Roll, Nonsterile, Absorbent, 1 lb roll
  • Cat. no.:
    470161-446
  • Supplier No.:
    79210006

Specifications

About this item

Quantity: 1 lb.

Customers Who Bought This Also Bought