ContactUsLinkComponent
SOIL NPK TEST KIT GLOBE LAMOTTE
LaMotte® NPK Soil Test Kit
# de catalogue 470157-410
Fournisseur:  LaMotte
Numéro CAS:  
SOIL NPK TEST KIT GLOBE LAMOTTE
LaMotte® NPK Soil Test Kit
# de catalogue 470157-410
Fournisseur:  LaMotte
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    LaMotte® NPK Soil Test Kit
  • Cat. no.:
    470157-410
  • Supplier No.:
    5880-6
  • No. of Tests:
    50
Spécifications
A propos de cet article
Safe, easy, economical test for three factors.
  • No hazardous compounds required for testing
  • Useful for the lab and field work
  • Includes instructions
  • Aligned with GLOBE protocols
Test for nitrogen, potassium, and phosphorus with one kit; the special TesTabs, nonhazardous reagents in tablet form, make it simple. The kit comes with testing apparatus, illustrated instructions, and a laminated color chart for comparison all packaged in a light carrying case for transportation to the field. You get enough materials for 50 tests each.
Customers Who Bought This Also Bought