Spécifications
- Description:LaMotte® NPK Soil Test Kit
- Cat. no.:470157-410
- Supplier No.:5880-6
- No. of Tests:50
Spécifications
A propos de cet article
Safe, easy, economical test for three factors.
- No hazardous compounds required for testing
- Useful for the lab and field work
- Includes instructions
- Aligned with GLOBE protocols
Test for nitrogen, potassium, and phosphorus with one kit; the special TesTabs, nonhazardous reagents in tablet form, make it simple. The kit comes with testing apparatus, illustrated instructions, and a laminated color chart for comparison all packaged in a light carrying case for transportation to the field. You get enough materials for 50 tests each.