Stainless Steel Spoon/Spatula
470149-044
- Description:Stainless steel spoon/spatula
- Cat. no.:470149-044
- Supplier No.:CS2519/3
Constructed entirely of stainless steel.
One end is spoon shaped, 13 mm wide, and the other is flat, with square corners (8 mm wide).
Customers Who Bought This Also Bought