Economy Mortar & Pestle Sets
# de catalogue 470148-802
Fournisseur: SILICO & CHEMICO PORCELAIN SE
Spécifications
- Description:Mortar & Pestle Set, 130 mm, 300 ml
- Cat. no.:470148-802
- Supplier No.:12091-254
Spécifications
A propos de cet article
Quickly grind materials to a powder.
- Glazed on the outside
- Pouring spout
Affordable student quality sets with pour-out lips. Designed for general laboratory use, the mortars are glazed on the outside and unglazed on the inside.
Customers Who Bought This Also Bought