Crucible Tongs
# de catalogue 470148-786
Fournisseur: VWR International
Spécifications
- Description:Stainless Steel
- Cat. no.:470148-786
- Supplier No.:CH13010-3
Spécifications
A propos de cet article
These tongs are designed for the safe handling of hot objects.
- Overall length is 9"
- Riveted joints and serrated tips
Pick up hot crucibles safely. The overall 9" length keeps you at a safe distance ion moving hot crucibles, all while maintaining a controlling grip.
Customers Who Bought This Also Bought