Economy Mortar & Pestle Sets
# de catalogue 470148-746
Fournisseur: SILICO & CHEMICO PORCELAIN SE
Spécifications
- Description:Mortar & Pestle Set, 100 mm, 100 ml
- Cat. no.:470148-746
- Supplier No.:12091-253
Spécifications
A propos de cet article
Quickly grind materials to a powder.
- Glazed on the outside
- Pouring spout
Affordable student quality sets with pour-out lips. Designed for general laboratory use, the mortars are glazed on the outside and unglazed on the inside.
Customers Who Bought This Also Bought