Funnel Set, Dynalon
# de catalogue 470146-840
Fournisseur: Dynalon
Spécifications
- Description:Funnel Set
- Cat. no.:470146-840
- Supplier No.:2416450SET
Spécifications
A propos de cet article
Two each of top diameter 65mm and 100mm and 120mm. All are 60° angle. Set of six.
Customers Who Bought This Also Bought