Tape Measure
Catalog # 470145-602
Supplier: Skywill Far East Industries
Specifications
- Description:Tape measure, metric and imperial
- Length:1.5 m (5 ft.)
- Cat. no.:470145-602
- Supplier No.:5006-6
Specifications
About this item
1.5m dual scale (inches and centimeters) tape measures.
- Dual scale
- Plastic with metal tabs
Measures are plastic with metal tabs at the ends.
Customers Who Bought This Also Bought