Order Entry
ContactUsLinkComponent
SCOOP 250ML 6.4CM HANDLE POLYPR PK12
Scoop, 250 ml, 6.4 cm Handle
# de catalogue 470145-134
Fournisseur:  Bel-Art
SCOOP 250ML 6.4CM HANDLE POLYPR PK12
Scoop, 250 ml, 6.4 cm Handle
# de catalogue 470145-134
Fournisseur:  Bel-Art

Spécifications

  • Description:
    Scoop, 250mL, 6.4cm Handle
  • Cat. no.:
    470145-134
  • Supplier No.:
    F36753-000

Spécifications

A propos de cet article

Rigid and strong scoop made from white polypropylene.

  • Flat bottom keeps it upright when filled or empty
  • Steam autoclavable at 121ºC (250ºF)

Designed for retrieving samples from a larger source.

Scoop, 250mL, 6.4cm Handle

Customers Who Bought This Also Bought