Luer Lock Stop Cocks
# de catalogue 470102-722
Fournisseur: S17 SCIENCE LLC.
Spécifications
- Description:Stopcock, 2-Way, Luer Lock, Male/ Female
- Cat. no.:470102-722
- Supplier No.:EQ 066
Spécifications
A propos de cet article
Assortment of 2-way stopcocks, syringes, and syringe extenders control the flow of liquids and gases, and make gas-tight connections.
- Polypropylene
Customers Who Bought This Also Bought