ContactUsLinkComponent
BUCHNER FUNNEL 70MM PP
Polypropylene Buchner Funnel
# de catalogue 470174-670
BUCHNER FUNNEL 70MM PP
Polypropylene Buchner Funnel
# de catalogue 470174-670
Spécifications
  • Description:
    Polypropylene Buchner Funnel
  • Cat. no.:
    470174-670
  • Supplier No.:
    57105
Spécifications
A propos de cet article
Useable With or Without Filter Paper.
  • Uses 7.0 cm diameter filter paper
Two-piece HDPE funnel with sealed-in, polyethylene filter plate of coarse porosity offers good retention and fast flow under vacuum.
Customers Who Bought This Also Bought