ContactUsLinkComponent
FUNNEL 70MM DIAMETER PK12
Economy Short Stem Funnels, Hutzler
# de catalogue 470150-352
Fournisseur:  Hutzler Manufacturing
FUNNEL 70MM DIAMETER PK12
Economy Short Stem Funnels, Hutzler
# de catalogue 470150-352
Fournisseur:  Hutzler Manufacturing
Spécifications
  • Description:
    Economy Short Stem Funnels, Top ID, mm: 70, Stem Length, mm: 19
  • Cat. no.:
    470150-352
  • Supplier No.:
    802
Spécifications
A propos de cet article
Funnels are made of durable plastic. Funnels of top inner diameter 70-108 are polypropylene and autoclavable. Funnels with top inner diameter of 127 and 165 are polyethylene and not autoclavable.
The 165 mm size has a wide stem for quick transfers.
Customers Who Bought This Also Bought