Economy Spatulas
# de catalogue 470149-446
Fournisseur: VWR International
Spécifications
- Description:Economy spatulas, Length: 6"
- Cat. no.:470149-446
- Blade Length:152 mm (6")
- Supplier No.:CS2518/1
Spécifications
A propos de cet article
Stainless steel spatulas, with one end flat and the other (rounded end) bent for easy scooping.
- Available in two sizes
Customers Who Bought This Also Bought