Stainless Steel Spoon/Spatula
Catalog # 470149-440
Supplier: VWR International
Specifications
- Description:Stainless steel spoon/spatula
- Cat. no.:470149-440
- Supplier No.:CS2519/1
Specifications
About this item
Constructed entirely of stainless steel.
One end is spoon shaped, 13 mm wide, and the other is flat, with square corners (8 mm wide).
Customers Who Bought This Also Bought