Scoop Type Spatula
# de catalogue 470149-438
Fournisseur: VWR International
Spécifications
- Description:Spatula scoop
- Cat. no.:470149-438
- Supplier No.:CS2524/3
Spécifications
A propos de cet article
This scoop-type spatula is versatile and durable.
- Spatula dimensions: ¹/₂×7¹/₂"
- One rounded end, one pointed end
The stainless steel spatula has a semicircular cross section.
Customers Who Bought This Also Bought