Lil’pet Micropipette
# de catalogue 470148-818
Fournisseur: VWR International
Spécifications
- Volume:500 µl
- Cat. no.:470148-818
- Supplier No.:MM-500
Spécifications
A propos de cet article
Lil’pet Micropipettes feature ideal volumes for gel loading.
- Units accept standard micropipette tips
- Economical
- Durable
- Autoclavable
These fixed-volume pipettes feature a double stop plunger to expel all liquid from the tip.
The 1000 μl model includes two tips. All other pipettes come with one each.
Customers Who Bought This Also Bought