Flint Glass Watch Glasses
Catalog # 470144-840
Supplier: United Scientific Supplies
Specifications
- Ø:125 mm
- Description:VWR® watch glass
- Cat. no.:470144-840
- Supplier No.:WGL125
Specifications
About this item
Great Affordable Labware
- Fire-polished edge
- All sizes molded to the same radius
The watches glasses can be stacked together.
Customers Who Bought This Also Bought