ContactUsLinkComponent
VWR WATCH GLASS 125MM - 5 5IN
Flint Glass Watch Glasses
Catalog # 470144-840
VWR WATCH GLASS 125MM - 5 5IN
Flint Glass Watch Glasses
Catalog # 470144-840
Specifications
  • Ø:
    125 mm
  • Description:
    VWR® watch glass
  • Cat. no.:
    470144-840
  • Supplier No.:
    WGL125
Specifications
About this item
Great Affordable Labware
  • Fire-polished edge
  • All sizes molded to the same radius
The watches glasses can be stacked together.
Customers Who Bought This Also Bought