Beaker Tri pour
# de catalogue 470142-552
Fournisseur: DYN-A-MED PRODUCTS
Spécifications
- Capacity:800 ml (27 oz.)
- Description:Beaker Tri Pour 800 mL, Pkg./10
- Cat. no.:470142-552
- Supplier No.:80097
Spécifications
A propos de cet article
This tri-pour beaker is perfect for any classroom. The 800ml beakers are extremely durable and portable. They make science easy and the polypropylene material is safe to use.
Customers Who Bought This Also Bought