ContactUsLinkComponent
NEULOG GSR LOGGER SENSOR
NeuLog Galvanic Skin Response (GSR) Logger Sensor
# de catalogue 470137-402
Fournisseur:  Wards
NEULOG GSR LOGGER SENSOR
NeuLog Galvanic Skin Response (GSR) Logger Sensor
# de catalogue 470137-402
Fournisseur:  Wards
Spécifications
  • Interface:
    USB
  • Resolution:
    10 nS
    ADC: 10 bit
  • Description:
    Neulog Galvanic skin response (GSR) logger sensor
  • Measurement time:
    1 s - 31 days
  • Cat. no.:
    470137-402
  • Range:
    0 - 10 μS
    Arbitrary units: 0 - 65279
  • Supplier No.:
    NUL-217
Spécifications
A propos de cet article
Measures the Psycho-Galvanic Reflex.
The Galvanic Skin Response (GSR) logger sensor measures the conductivity of our skin as it changes according to unconscious emotion effects such as sudden noise, smell, touch, pain or view. This sensor has two ranges: conductivity in micro Siemens and arbitrary numbers. It also supports experiments lasting between 1 second and 31 days.
For each probe you will need one or more of the following items: Battery Module, Viewer module(Not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
  • Max. sample rate 100 S/sec.
The Galvanic Skin Response (GSR) logger sensor measures the conductivity of our skin as it changes according to unconscious emotion effects such as sudden noise, smell, touch, pain or view. This sensor has two ranges: conductivity in micro Siemens and arbitrary numbers.
Ordering information: For each probe you will need one or more of the following items: Battery Module, Viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.
Customers Who Bought This Also Bought