Spécifications
- Resolution:100 µs
- Description:NeuLog photogate logger sensor
- Cat. no.:470137-386
- Accuracy:100 µs
- Supplier No.:NUL-209
Spécifications
A propos de cet article
This sensor measures acceleration with one or two photogates and shows the status pictorially.
Study motion via four modes of operation: time, velocity, momentum or acceleration. Easily measure acceleration with one or two photogates and associated timing cards and show the status pictorially. Select the modes of operation by simply clicking on their icon. Displays values either digitally or graphically.
For each probe you will need one or more of the following items: Battery Module, Viewer module(Not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
- Simple display features
- Four modes of operation: time, velocity, momentum, or acceleration
- Sensor displays values digitally and graphically
- Ideal for researching motion
Study motion via four modes of operation: time, velocity, momentum, or acceleration. You can select the modes of operation by clicking on their icon.
The sensor displays values digitally or graphically.