VWR® Pipette Fillers
# de catalogue 470123-222
Fournisseur: Heathrow Scientific
Spécifications
- Accuracy:±0.01 ml
- Cat. no.:470123-222
- Supplier No.:HS20632A
Spécifications
A propos de cet article
Pipette fillers eliminate the hazardous practice of drawing liquids into pipettes by mouth.
- Filters work with pipettes up to 50 ml in size
- Filters help avoid sample contamination
- Safe, precise delivery
- Ideal for volatile, corrosive, or poisonous liquids
- Fits a standard serological pipet
To use, insert the pipette into the bottom opening of the filler, draw to the desired volume, and expel by pressing lettered valves.
The filters are made of soft rubber. Colors may vary. Dimensions: 54×54×130 mm (L×W×D).
Customers Who Bought This Also Bought