Tape Measure
Catalog # 470122-698
Supplier: VWR International
Specifications
- Description:Tape measure, metric and imperial
- Length:3 m (10 ft.)
- Cat. no.:470122-698
- Supplier No.:PH0074B
Specifications
About this item
Bright yellow steel tape is marked in both English and metric. Protected by a high impact plastic case and includes a belt clip, loop handle and automatic rewind.
- Length: 3 m (10')
Customers Who Bought This Also Bought