ContactUsLinkComponent
BURNER TIPS - SPARK METAL PK 5
Burner Lighters with Spring-Action
Catalog # 470120-594
BURNER TIPS - SPARK METAL PK 5
Burner Lighters with Spring-Action
Catalog # 470120-594
Specifications
  • Description:
    Spark tips with 1500 ignitions
  • Cat. no.:
    470120-594
  • Supplier No.:
    GSFLT1PK4
Specifications
About this item
Use for lghting gas burners.
  • Flat file set in large shield to trap gas for quick ignition
  • Replacement tips available
The hooded lighter allows you to safety light all variety of gas burners.
Customers Who Bought This Also Bought