Burner Lighters with Spring-Action
Catalog # 470120-594
Supplier: United Scientific Supplies
Specifications
- Description:Spark tips with 1500 ignitions
- Cat. no.:470120-594
- Supplier No.:GSFLT1PK4
Specifications
About this item
Use for lghting gas burners.
- Flat file set in large shield to trap gas for quick ignition
- Replacement tips available
The hooded lighter allows you to safety light all variety of gas burners.
Customers Who Bought This Also Bought