Burner Lighters with Spring-Action
# de catalogue 470120-592
Fournisseur: United Scientific Supplies
Spécifications
- Description:Burner lighter with spring-action, 8"
- Supplier No.:GLST01
Spécifications
A propos de cet article
Use for lghting gas burners.
- Flat file set in large shield to trap gas for quick ignition
- Replacement tips available
The hooded lighter allows you to safety light all variety of gas burners.
Customers Who Bought This Also Bought