ContactUsLinkComponent
BURNER LIGHTER - SPRING ACTION
Burner Lighters with Spring-Action
# de catalogue 470120-592
BURNER LIGHTER - SPRING ACTION
Burner Lighters with Spring-Action
# de catalogue 470120-592
Spécifications
  • Description:
    Burner lighter with spring-action, 8"
  • Supplier No.:
    GLST01
Spécifications
A propos de cet article
Use for lghting gas burners.
  • Flat file set in large shield to trap gas for quick ignition
  • Replacement tips available
The hooded lighter allows you to safety light all variety of gas burners.
Customers Who Bought This Also Bought