ContactUsLinkComponent
SIM PARTS ARTIFICIAL BLOOD 1 LITER
Artificial Blood For Simulators
# de catalogue 470119-456
Fournisseur:  Nasco
SIM PARTS ARTIFICIAL BLOOD 1 LITER
Artificial Blood For Simulators
# de catalogue 470119-456
Fournisseur:  Nasco
Spécifications
  • Description:
    Artificial Blood Mix, 1L
  • Cat. no.:
    470119-456
  • Supplier No.:
    W44061
Spécifications
A propos de cet article
For Use In Life/Form Iv Simulators
  • Adds a sense of reality to simulator practice
  • Easy to flush from the simulator when finished
Although not the same viscosity as real blood, Life/form® simulation blood provides a sense of reality to any activity. For use with Life/form® trainers.
Customers Who Bought This Also Bought