Porous Cups
# de catalogue 470104-080
Fournisseur: VWR International
Spécifications
- Description:Porous cups
- Cat. no.:470104-080
- Diameter ×H:36×72 mm
- Supplier No.:PRCUP
Spécifications
A propos de cet article
Porous cylindrical cups.
- Made of ceramic
Pore diameter is approximately 0.5 µm.
Customers Who Bought This Also Bought