ContactUsLinkComponent
TANK WATER WEATHER WATER TANK 4702001
Weather Water Tanks: Fronts and Thermoclines Lab Activity
# de catalogue 470042-990
Fournisseur:  CURBELL PLASTICS
TANK WATER WEATHER WATER TANK 4702001
Weather Water Tanks: Fronts and Thermoclines Lab Activity
# de catalogue 470042-990
Fournisseur:  CURBELL PLASTICS
Spécifications
  • Description:
    Single Weather Water Tank Only
  • Cat. no.:
    470042-990
  • Supplier No.:
    486900
Spécifications
A propos de cet article
What happens when warm ocean water and cold ocean water meet?
This easy-to-perform, hands-on lab is a great way for students to see (and remember) how fluids of different temperatures and densities interact, making it a useful tool for the study of weather (fronts) and oceanography (thermoclines). This activity simulates these phenomena.
In the first part of the activity, each group of students places differently colored hot and cold water into a divided tank. When they remove the barrier, students are amazed to see the cold water sliding below the hot water while the hot water moves up and over the cold water. As they continue the activity, they find that not only do hot and cold water form distinct layers, but adding warm water to these layers results in a third layer between the first two. This activity employs the Framework Science Standard ESS2: Earth's Systmes.
This kit has been aligned with all published National Standards. Pre- and Post-laboratory assessments and vocabulary words all target specific Science and Engineering Practices and common core standards. The teacher guide also contains specific "Did You Know" concepts that link the real world with the scientific standards explored. A student copymaster is also included. This kit contains enough materials for six groups of 4-5 students.
Materials included:
6 Water tanks
1 Blue food coloring, 30 mL
1 Red food coloring, 20 mL
Student Guide and Teacher's Guide
Materials not included:
14 Disposable pipets or droppers
12 Beakers, 400 mL or 500 mL
Hot and cold water
Sheets of unlined white paper or cardboard
Tape
Time requirements:
This lab activity takes approximately 45 minutes to complete
  • Students Research Weather and Oceanography Concepts Material for Six Groups Kit Employs Framework Science Standard ESS2 Estimated Lab Time: 45 Minutes Assessments and Vocabulary Words Target Common Core Standards
This easy-to-perform, hands-on lab is a great way for students to see and remember how fluids of different temperatures and densities interact. This activity simulates weather fronts and oceanography (thermoclines), making it a useful tool for the study of these phenomena.
In the first part of the activity, each group of students places two different colors of hot and cold water into a divided tank. When they remove the barrier, students are amazed to see the cold water sliding below the hot water while the hot water moves up and over the cold water. As students continue the activity, they find that not only do hot and cold water form distinct layers but also that adding warm water to these layers results in a third layer between the first two. This activity employs the Framework Science Standard ESS2: Earth's Systems. It is aligned with all published National Standards. Pre and post-laboratory assessments and vocabulary words target specific Science and Engineering Practices and Common Core standards. The teacher guide also contains specific "Did You Know" concepts that link the real world with the scientific standards explored. This kit contains a student copymaster and enough materials for six groups of four to five students. This lab activity takes approximately 45 minutes to complete.
Ordering information: This kit includes six water tanks, 30 mL of blue food coloring, 20 mL of red food coloring, a student's guide, and a teacher's guide. 14 disposable pipets or droppers, 12 beakers (400 or 500 mL), hot and cold water, cardboard or sheets of unlined white paper, and tape are required but not included.
Customers Who Bought This Also Bought