Quick-Developing Rotifers
Catalog # 470030-310
Supplier: FLORIDA AQUA FARMS INC
Specifications
- Description:Quick-Developing Rotifers
- Cat. no.:470030-310
- Supplier No.:873120
Specifications
About this item
Rotifers are well adapted to adverse conditions; when dry, the eggs remain viable for years.
- Mixed species
Now you can demonstrate this adaptation by reconstituting the dry eggs supplied. Just add water, and the eggs of different Rotifer species begin to hatch in 24 hours, completing their process in only 72 hours.
These freshwater rotifers grow best on a diet of microalgae and Roti-Rich, available separately. Instructions are included.
Ordering information: This item will ship immediately upon order, unless you specify otherwise.
Delivery: This item ships directly from our supplier.
Customers Who Bought This Also Bought