Glass Stirring Rods
# de catalogue 470020-786
Fournisseur: United Scientific Supplies
Spécifications
- Ø:5 mm
- Length:150 mm
- Cat. no.:470020-786
- Supplier No.:GSR006
Spécifications
A propos de cet article
Glass stirring rod features fire polished flint glass.
- Rounded ends
Stirring rods are made of soft flint glass, with both ends fire-polished and rounded.
Customers Who Bought This Also Bought