Fine-Tip Brushes
# de catalogue 470020-022
Fournisseur: SOLO HORTON BRUSHES, INC.
Spécifications
- Bristle Material:Camel hair
- Description:Fine-Tip Brushes
- Total length:17.8 cm (7")
- Cat. no.:470020-022
- Supplier No.:2115-2
Spécifications
A propos de cet article
The soft, natural camel hair bristles of this fine tipped brush will not harm delicate structures like stiff forceps or needles will.
Customers Who Bought This Also Bought