Order Entry
ContactUsLinkComponent
SLOTTED WT HNGR 50G SILVER COLOR
Mass Hangers
# de catalogue 470017-096
Fournisseur:  VWR International
SLOTTED WT HNGR 50G SILVER COLOR
Mass Hangers
# de catalogue 470017-096
Fournisseur:  VWR International

Spécifications

  • Description:
    Mass Hanger, 1500 g Capacity
  • Cat. no.:
    470017-096
  • Weight:
    50 g
  • Supplier No.:
    PH0258-HNG

Spécifications

A propos de cet article

Replacement hanger for slotted masses.

A top hook and precise weight make this stainless steel hanger ideal for use with slotted weights

Customers Who Bought This Also Bought