Student Grade Economy Microscope Slides
Catalog # 470016-270
Supplier: VWR International
Specifications
- Description:Plain glass slides, soda lime glas
- L×W:3×1"
- Thickness:1.0 - 1.2 mm
- Cat. no.:470016-270
- Supplier No.:BI0082/PK72
Specifications
About this item
These economical slides are available in a variety of types.
- Various Slide Types for All Needs
- Standard Slide Size
- Optically Clear Glass Construction
- Ground-Glass Edges
They’re perfect for consistently reliable mounts without the high cost of premium grade slides.
The slides are made of optically clear glass with ground-glass edges.
Customers Who Bought This Also Bought