Mass Hangers
# de catalogue 470016-120
Fournisseur: Wards
Spécifications
- Description:Mass Hanger, 500 g Capacity
- Cat. no.:470016-120
- Weight:5 g
- Supplier No.:9611-000
Spécifications
A propos de cet article
Replacement hanger for slotted masses.
A top hook and precise weight make this stainless steel hanger ideal for use with slotted weights
Customers Who Bought This Also Bought