ContactUsLinkComponent
PH PAPER WIDE RANGE VL100
Wide Range pH Test Strips
# de catalogue 470008-014
PH PAPER WIDE RANGE VL100
Wide Range pH Test Strips
# de catalogue 470008-014
Spécifications
  • Description:
    pH Paper, wide range, 2×¼", Vial/100
  • Supplier No.:
    7-3000-30
Spécifications
A propos de cet article
Ideal for classroom use, this general purpose pH paper provides a distinct color match for each pH unit from 1 to 14.
  • Convenient storage case
  • Easy to read color code
  • Aligned with GLOBE protocols
100 strips, measuring 2" long x 1/4" wide, are packaged in a plastic container along with a pH color matching chart.
Ordering information: Each container includes 5 books of 20 strips.
Customers Who Bought This Also Bought