Order Entry
ContactUsLinkComponent
FORCEPS HALSTEAD MOSQUITO SS 127 MM
Halstead Artery Forceps
Catalog # 470007-668
FORCEPS HALSTEAD MOSQUITO SS 127 MM
Halstead Artery Forceps
Catalog # 470007-668

Specifications

  • Description:
    Artery Forceps
  • Length:
    128 mm (5")
  • Material:
    Stainless steel
  • Cat. no.:
    470007-668
  • Tip Style:
    Straight
  • Supplier No.:
    140551

Specifications

About this item

Scissor handles securely clamp an injection needle to the vessel, preventing leaks. They have straight, serrated jaws and a box joint.

Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Customers Who Bought This Also Bought