ContactUsLinkComponent
SPOT-PLATE CLR PLSTY 15DPR 140X80MM PK10
Clear Polystyrene Spot Plates
# de catalogue 470005-972
Clear Polystyrene Spot Plates
# de catalogue 470005-972
Spécifications
  • Description:
    Clear Polystyrene Spot Plates
  • Cat. no.:
    470005-972
  • Supplier No.:
    A-M 9000
Spécifications
A propos de cet article
Offering more depressions than other spot plates, these plates are suitable for most lab applications. Made of clear polystyrene with an attached measuring scoop.
  • 15 depressions measure 2 ml deep
  • Each plate measures 140×80 mm with 15 depressions
Packaging: Package of 10.
Caution: Not recommended for use with strong solvents.
Customers Who Bought This Also Bought