Order Entry
ContactUsLinkComponent
TONGS FLASK/TEST TUBE 20 CM LONG
Specimen Tongs
# de catalogue 470005-898
Fournisseur:  Dunrite Instruments
TONGS FLASK/TEST TUBE 20 CM LONG
Specimen Tongs
# de catalogue 470005-898
Fournisseur:  Dunrite Instruments

Spécifications

  • Description:
    Tongs, Flask & Test Tube, 20 cm long
  • Cat. no.:
    470005-898
  • Supplier No.:
    6343500

Spécifications

A propos de cet article

Ideal for handling preserved specimens, the wide clamping jaws are flattened and have serrations to prevent slipping.

The jaw spread is 2 inches. The tongs are constructed of nickel-plated steel and have an overall length of 8 inches.

Customers Who Bought This Also Bought