Ward's® Bacti-Spreader
# de catalogue 470004-860
Fournisseur: Dunrite Instruments
Spécifications
- Length:9"
- Description:Bacti-Spreader
- Cat. no.:470004-860
- Blade Width:2"
- Supplier No.:151993
Spécifications
A propos de cet article
Special Design Evenly Distributes Bacteria
- Reusable for multiple inoculations
The stainless steel rod has been specifically designed for greater contact with agar for more even distribution of bacteria.
Customers Who Bought This Also Bought