ContactUsLinkComponent
ARSENIC QUICK TEST KIT 100 TESTS
Arsenic Quick Test Kit
# de catalogue 470021-304
ARSENIC QUICK TEST KIT 100 TESTS
Arsenic Quick Test Kit
# de catalogue 470021-304
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Arsenic Quick Test Kit
  • Cat. no.:
    470021-304
  • Supplier No.:
    481396
Spécifications
A propos de cet article
Safe, Accurate, and Cost-Effective Test for Arsenic in Water or Soil
This colorimetric arsenic test only takes 12 minutes to complete, and it will detect a range of 0–500 ppb. The kit comes with 100 tests.
Customers Who Bought This Also Bought