Single Action Stopwatch
Catalog # 470018-448
Supplier: MARATHON MANAGEMENT CO.
Specifications
- Description:Stopwatch, single action, 1/5 second, 30 minutes
- Cat. no.:470018-448
- Supplier No.:211003-B
Specifications
About this item
An excellent heavy-duty stopwatch that is ideal for student use.
- The stopwatch can be read to ¹/₅ second and registers to 30 minutes
- Attractive metal case
Operation is simple with start, stop, and reset controlled by successive crown depressions.
Customers Who Bought This Also Bought