Order Entry
ContactUsLinkComponent
MULT RL TAPE DISP-1 CORE UP TO 16 RLS
Multiple Roll Tape Dispenser, 1" Core
# de catalogue 470018-038
Fournisseur:  NEVS INK INC SE
MULT RL TAPE DISP-1 CORE UP TO 16 RLS
Multiple Roll Tape Dispenser, 1" Core
# de catalogue 470018-038
Fournisseur:  NEVS INK INC SE

Spécifications

  • Description:
    Multiple roll tape dispenser steel 1" Core
  • Cat. no.:
    470018-038
  • Supplier No.:
    VWR-EDU-PT

Spécifications

A propos de cet article

Designed for tape with 1" cores, this dispenser accommodates 1/2", 3/4", 1", 1 1/2", or 2" wide tapes for a total dispensing width of 9 1/2". Dispenser also features bottom grips to stay securely in place during use.

Customers Who Bought This Also Bought