Funnel Support with Base
# de catalogue 470005-858
Fournisseur: VWR International
Spécifications
- Dimensions:11 x 38 cm (Base)
- Description:Funnel Support with Base
- Height:43 cm
- Cat. no.:470005-858
- Supplier No.:CH0689B
Spécifications
A propos de cet article
Made from fine, oil finished hardwood designed to support up to four funnels. The height is adjustable and the holes are slotted for easy placement of funnels. The base measures 11 x 38 cm.
Customers Who Bought This Also Bought