Chromatography Paper
Catalog # 470003-724
Supplier: AHLSTROM MUNKSJO FILTRATION LLC.
Specifications
- Flow rate:130 mm/30 min
- Size:46 x 57 cm (18 x 22")
- Cat. no.:470003-724
- Supplier No.:6130-4657
Specifications
About this item
Quality paper for general chromatography and electrophoresis.
- White, smooth sheets with a hard surface
Customers Who Bought This Also Bought