ContactUsLinkComponent
Tip,200R Barrier large cs5Packs
ART® REACH™ Pipette Tips, Thermo Scientific
# de catalogue 89413-262
Fournisseur:  Thermo Scientific
Tip,200R Barrier large cs5Packs
ART® REACH™ Pipette Tips, Thermo Scientific
# de catalogue 89413-262
Fournisseur:  Thermo Scientific
Spécifications
  • Length:
    9.09 cm (3½")
  • Volume:
    20 - 200 µL
  • Packed:
    96/Rack
  • Sterile:
    +
  • Tip Style:
    ART® 200 REACH™
  • Supplier No.:
    2160P-05-HR
Spécifications
A propos de cet article
Available in two styles, the ART® REACH™ barrier tip, with a self-sealing barrier to eliminate carryover contamination, and the ART® REACH™ non-filter tip, without a barrier when cross-contamination is not a concern.
Ordering information: Available in traditional lift-off racks and versatile hinged racks. Low retention tips 89413-262, -314, and -326. Tips 89413-324 and -326 are specifically designed for Gilson Pipetman and Thermo Scientific Matrix pipettes.
Customers Who Bought This Also Bought