Spécifications
- Dimensions:15.4×14.3×5.6H cm (6×5.7×2.2”) with hinges and tabs; 14.3×14.3×5.6H cm (5.7×5.7×2.2”) without hinges and tabs
- Color:Natural
- Cat. no.:30621-380
- Supplier No.:F18851-0011
Spécifications
A propos de cet article
100-place polypropylene storage boxes provide compact, safe storage for 0.5, 1.5 and 2.0 mL tubes and cryo vials.
- Polypropylene construction
- Available in a variety of color choices
- Hinged lids with snap tabs
- Alphanumeric labeling
Great for organization on the bench, in the freezer, or during transport, they can be used at temperatures –80°C to +121°C (112°F to +249°F)
Hinged lids with snap tab open easily and close securely, while clear translucent color always lets you see inside. Opaque black boxes are ideal for light sensitive samples. Each box position is labeled with an alphanumeric grid for easy management of individual samples. ‘No slide’ features molded in bottom and lid provide secure stacking.
Unlike cardboard boxes they will not break down if exposed to moisture or heavy use. Five color choices let you visually sort and organize samples of different types.